Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Cyclohexylethanol 95%

Supplier: Ambeed
Warning

A498693-5G A498693-25G A498693-1G
77265-650EA 11 USD InStock
77265-650 77265-646 77265-648
2-Cyclohexylethanol 95%
2-Cyclohexylethanol
Caution: for Research Use Only


Formula: C₈H₁₆O
MW: 128.21 g/mol
MDL Number: MFCD00001525
CAS Number: 4442-79-9
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty