Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-(Methylthio)phenol 98%

Supplier: Ambeed
Warning
Synonyms: 2-Hydroxythioanisole

A527117-5G A527117-1G A527117-25G
77265-964EA 12 USD InStock
77265-964 77265-966 77265-968
2-(Methylthio)phenol 98%
2-(Methylthio)phenol
Caution: for Research Use Only


Formula: C₇H₈OS
MW: 140.21 g/mol
MDL Number: MFCD00002211
CAS Number: 1073-29-6
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty