Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

cis-Dichlorobis(triethylphosphine)platinum(II) 98%

Supplier: Ambeed
Warning

A552715-250MG A552715-1G A552715-100MG
77253-850EA 372 USD InStock
77253-850 77253-852 77253-848
cis-Dichlorobis(triethylphosphine)platinum(II) 98%
cis-Dichlorobis(triethylphosphine)platinum(II)
Caution: for Research Use Only


Formula: PtCl₂[P(C₂H₅)₃]₂
MDL Number: MFCD00058821
CAS Number: 15692-07-6
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty