Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4,4',4''-Trimethyltrityl alcohol 98%

Supplier: Ambeed
Warning
Synonyms: Tri-p-tolylmethanol

A557820-250MG A557820-5G A557820-1G
77292-488EA 49 USD InStock
77292-488 77292-492 77292-490
4,4',4''-Trimethyltrityl alcohol 98%
4,4',4''-Trimethyltrityl alcohol
Caution: for Research Use Only


Formula: C₂₂H₂₂O
MW: 302.42 g/mol
MDL Number: MFCD00014919
CAS Number: 3247-00-5
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty