Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Thiophenecarbonitrile 98%

Supplier: Ambeed
Danger
Synonyms: Thiophene-2-carbonitrile2-Thienyl cyanide2-Cyanothiophene

A560039-5G A560039-10G A560039-25G
77292-514EA 10 USD InStock
77292-514 77292-518 77292-516
2-Thiophenecarbonitrile 98%
2-Thiophenecarbonitrile
Caution: for Research Use Only


Formula: C₅H₃NS
MW: 109.15 g/mol
MDL Number: MFCD00005416
CAS Number: 1003-31-2
UN: 2924
ADR: 3,8,Ⅲ
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty