Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3,5-Dichloro-4-fluorobenzoic acid 98%

Supplier: Ambeed
Warning

A570305-100G A570305-25G A570305-5G A570305-1G
77280-098EA 113 USD InStock
77280-098 77280-100 77280-094 77280-096
3,5-Dichloro-4-fluorobenzoic acid 98%
3,5-Dichloro-4-fluorobenzoic acid
Caution: for Research Use Only


Formula: C₇H₃Cl₂FO₂
MW: 209.0019 g/mol
MDL Number: MFCD03840463
CAS Number: 98191-30-1
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty