Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Mercaptopropyl(dimethoxy)methylsilane 97% (by GC)

Supplier: Ambeed
Danger
Synonyms: 3-Mercaptopropylmethyldimethoxysilane3-(Dimethoxymethylsilyl)-1-propanethiol

A573784-25G
77280-124EA 10 USD InStock
77280-124
3-Mercaptopropyl(dimethoxy)methylsilane 97% (by GC)
3-Mercaptopropyl(dimethoxy)methylsilane
Caution: for Research Use Only


Formula: C₆H₁₆O₂SSi
MW: 180.34 g/mol
MDL Number: MFCD00053923
CAS Number: 31001-77-1
UN: 3334
ADR: 9,Ⅲ
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty