Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Fluoro-4-nitrobenzene 98%

Supplier: Ambeed
Danger
Synonyms: 4-Fluoronitrobenzene1-Fluoro-4-nitrobenzene

A587794-25G A587794-10G
77292-686EA 6 USD InStock
77292-686 77292-684
1-Fluoro-4-nitrobenzene 98%
1-Fluoro-4-nitrobenzene
Caution: for Research Use Only


Formula: C₆H₄FNO₂
MW: 141.1 g/mol
MDL Number: MFCD00007282
CAS Number: 350-46-9
UN: 2811
ADR: 6.1,Ⅲ
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty