Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3,5-Dichloro-2-hydroxybenzenesulfonic acid sodium salt 98%

Supplier: Ambeed
Warning
Synonyms: Sodium 3,5-Dichloro-2-hydroxybenzenesulphonateSodium 3,5-Dichloro-2-hydroxybenzenesulfonate

A601983-100G A601983-25G A601983-5G
77280-246EA 113 USD InStock
77280-246 77280-248 77280-250
3,5-Dichloro-2-hydroxybenzenesulfonic acid sodium salt 98%
3,5-Dichloro-2-hydroxybenzenesulphonic acid sodium salt
Caution: for Research Use Only


Formula: C₆H₃Cl₂NaO₄S
MW: 265.05 g/mol
MDL Number: MFCD00009798
CAS Number: 54970-72-8
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty