Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Aminophthalide 98%

Supplier: Ambeed
Warning

A623409-250MG A623409-100MG A623409-25G A623409-1G A623409-5G
77242-824EA 273 USD InStock
77242-824 77242-820 77242-822 77242-816 77242-818
5-Aminophthalide 98%
5-Aminophthalide
Caution: for Research Use Only


Formula: C₈H₇NO₂
MDL Number: MFCD00778315
CAS Number: 65399-05-5
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty