Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4,4'-Difluorobiphenyl 98%

Supplier: Ambeed
Danger
Synonyms: PFB No. 154,4-Difluoro-1,1-biphenyl

A687762-5G A687762-25G A687762-1G
77293-184EA 125 USD InStock
77293-184 77293-186 77293-182
4,4'-Difluorobiphenyl 98%
4,4'-Difluorobiphenyl
Caution: for Research Use Only


Formula: C₁₂H₈F₂
MW: 190.19 g/mol
MDL Number: MFCD00000349
CAS Number: 398-23-2
UN: 3077
ADR: 9,Ⅲ
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty