Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Butoxybenzoyl chloride 99%

Supplier: Ambeed
Danger
Synonyms: 4-N-butoxybenzoylchloride

A697579-250MG A697579-1G
77293-258EA 17 USD InStock
77293-258 77293-256
4-Butoxybenzoyl chloride 99%
4-Butoxybenzoyl chloride
Caution: for Research Use Only


Formula: C₁₁H₁₃ClO₂
MW: 212.68 g/mol
MDL Number: MFCD00000689
CAS Number: 33863-86-4
UN: 3265
ADR: 8,Ⅱ
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty