Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Bromobutyryl chloride 95%

Supplier: Ambeed
Danger

A708194-1G A708194-250MG A708194-25G A708194-5G
77293-314EA 12 USD InStock
77293-314 77293-310 77293-312
4-Bromobutyryl chloride 95%
4-Bromobutyryl chloride
Caution: for Research Use Only


Formula: C₄H₆BrClO
MDL Number: MFCD00000753
CAS Number: 927-58-2
UN: 3265
ADR: 8,Ⅱ
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty