Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Nitrobenzonitrile 98%

Supplier: Ambeed
Warning
Synonyms: o-Nitrobenzonitrile

A757779-25G A757779-5G A757779-100G
77281-180EA 194 USD InStock
77281-180 77281-184 77281-182
2-Nitrobenzonitrile 98%
2-Nitrobenzonitrile
Caution: for Research Use Only


Formula: C₇H₄N₂O₂
MW: 148.12 g/mol
MDL Number: MFCD00007044
CAS Number: 612-24-8
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty