Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Chloro-2-iodoethane 97% stabilized

Supplier: Ambeed
Danger

Stabilization: Stabilized with Copper chip

A828123-1G
77244-360EA 24 USD InStock
77244-360
1-Chloro-2-iodoethane 97% stabilized
1-Chloro-2-iodoethane
Caution: for Research Use Only


Formula: C₂H₄ClI
MW: 190.41 g/mol
MDL Number: MFCD00041453
CAS Number: 624-70-4
UN: 1993
ADR: 3,Ⅲ
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty