Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Methylimidazole 98%

Supplier: Ambeed
Danger
Synonyms: 2-Methyl-1H-imidazole

A868819-500G A868819-25G A868819-100G A868819-5G
77281-786EA 5 USD InStock
77281-786 77281-784 77281-782 77281-780
2-Methylimidazole 98%
2-Methylimidazole
Caution: for Research Use Only


Formula: C₄H₆N₂
MW: 82.11 g/mol
MDL Number: MFCD00005190
CAS Number: 693-98-1
UN: 3263
ADR: 8,Ⅱ
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty