Would you like to log in again?
PYROSTAR™ ES-F LAL Reagents
Supplier: Fujifilm Wako Chemicals USA
WPEPK4-50015 WPEM-20006 WPEK4-20125 WPEK4-20003 WPEK4-20025 WPEK4-50015 WPEM-20025 WPEM-50015 WPEM-20015 WPEPK4-20015 WPESK-0025 WPEM-20125 WPESK-0003 WPESK-0015 WPEK4-20015 WPESK-0125 WPEPM-20015 WPEK4-50025 WPEK4-20006 WPEM-20003 WPEK4-50125 WPEPM-50015 WPEK4-50006 WPEK4-50003 WPEM-50025 WPEM-50003 WPEM-50125 WPEM-50006
76994-416EA 3342.72 USD InStock
76994-416 76994-418 76994-422 76994-400 76994-324 76994-424 76994-386 76994-342 76994-420 76994-344 76994-362 76994-384 76994-380 76994-408 76994-404 76994-412 76994-358 76994-336 76994-414 76994-374 76994-352 76994-398 76994-332 76994-410 76994-370 76994-350 76994-372 76994-394
PYROSTAR™ ES-F LAL Reagents
Microbiology Test Systems Endotoxin Tests
The PYROSTAR™ ES-F series of reagents are endotoxin-specific and specially formulated to be unreactive to (1,3)-ß-D-glucan. In addition, these products are dual-purpose, meaning they are formulated to be used as either a gel clot or kinetic turbidimetric assay. They are available in a single-test or multi-test configuration.
  • Endotoxin-specific reagent avoids false positive results from glucans
  • Dual-purpose reagent can perform either a gel clot or kinetic turbidimetric assay (KTA)
  • Gel clot sensitivities range from 0.015 to 0.25 EU/ml
  • Kinetic turbidimetric sensitivities range from 0.001 to 10 EU/ml
  • Due to its high sensitivity, chances of interference are reduced
  • Facilitates easier transition from gel clot to KTA
These reagents require only one Certificate of Analysis (COA), regardless of whether they are used as a gel clot or kinetic turbidimetric assay. PYROSTAR™ ES-F/Plate requires a different COA. All reagent kits are matched with a specific Control Standard Endotoxin (CSE). These reagents work well with samples that have color or with samples that present difficulties in quantification by the KCA.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty