Would you like to log in again?
PYROSTAR™ Neo
Supplier: Fujifilm Wako Chemicals USA
294-36731
76994-340EA 285.01 USD InStock
76994-340
PYROSTAR™ Neo
Microbiology Test Systems Endotoxin Tests
PYROSTAR™ Neo is a new line of endotoxin detection reagents developed by recombinant technology. It mimics traditional LAL reagents with three recombinant factors (Factor C, Factor B, and proclotting enzyme) and the pNA chromogenic group. Offerings an assay with high sensitivity and endotoxin specificity.
  • Colorimetric method, can be used with an absorbance plate reader
  • 3-Factor system mimics the same cascade reaction as traditional LAL
  • Endotoxin-specific reagent eliminates the risk of false positives from (1-3)ß-D-Glucan
  • synthetic and sustainable product
  • Quantitative range: 0.001 to 50 EU/ml
  • High sensitivity with less lot-to-lot variation
  • Stable storage after dissolution (4 h at 2 to 8 °C and 2 weeks at −30 °C)
PYROSTAR™ Neo is based on a genetically engineered approach to produce reactive factors required for endotoxin detection at pharmaceutical and medical equipment manufacturing sites. It also facilitates the same sensitivity in endotoxin detection as our limulus amebocyte lysate (LAL) reagent, PYROSTAR™ ES-F. This reagent utilizes the Kinetic Chromogenic Assay (KCA) to measure endotoxin content. It is composed of recombinant reactive factors equivalent to Factor C, Factor B and a proclotting enzyme, which constitute the horseshoe crab’s natural endotoxin reaction system, allowing for endotoxin detection that offers the same sensitivity as the LAL reagent. In addition, the absence of factor G eliminates interference due to reaction with β-glucan, enabling improved specificity.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty