Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5427R Centrifuges with Hydrocarbon Cooling, 230 V

Supplier: Eppendorf
5429000010 5429000010
76637-330EA 9521.85 USD InStock
CA76637-330 76637-330
5427R Centrifuges with Hydrocarbon Cooling, 230 V
Eppendorf Centrifuge 5427 R supports your greener laboratory with hydrocarbon cooling. It is the perfect solution for laboratories with a variety of molecular biology applications or those in need of high-throughput solutions for 1.5 to 2 ml tubes - all combined in a microcentrifuge with the highest throughput/footprint-ratio within the Eppendorf microcentrifuge range.

  • 230 V model
  • Natural refrigerant (R 290, Propane) means significantly reduced GWP (Global Warming Potential)
  • High RCF of 25001×g
  • FastTemp Function for rapid centrifuge pre-cooling, e.g. From 23 to 4 °C in approx. 11 min
  • ECO – shut off: Automatic compressor shut off for lower energy consumption when not in use
  • Highest throughput/footprint ratio of all Eppendorf microcentrifuges
  • Eppendorf QuickLock® rotor lid for all aerosol-tight rotors
  • Nine different rotors

Eppendorf Centrifuge 5427 R is a high-speed, refrigerated microcentrifuge for high-throughput research. It doubles the tube capacity with its 48-place double-row rotor, while maintaining safety and reliability.

Certifications: ACT certified.

Caution: 230 V model. Corresponding models with voltage suitable for use in US (76637-332, 76637-334, 76637-336) are also available, please see separate product page.

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty