Would you like to log in again?
AquaSprouts Aqua Plugs
Supplier: AQUASPROUTS, INC
AquaSprouts
7999
470353-522CT 60 USD InStock
470353-522
AquaSprouts Aqua Plugs
Animal Supplies Animal Environmental Enrichment
High quality peat and organic amendments create the perfect support for seed germination and cuttings.
  • Consistent moisture control
  • Infused micronutrients and biologicals
  • Porous to maintain aeration
  • Easy transplanting and handling
  • Compatible with most hydroponic and aquaponic grow systems
Special formula grow plugs anchor and nourish seedlings and small cuttings. Active biologicals and micronutrients create a perfect environment for young plants, to boost germination and root growth. Essential for starting small seeds in the AquaSprouts clay pebble medium or other pebble media, and great for transplanting.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty