Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Glycolic acid 98%

Supplier: Ambeed
Danger
Synonyms: 2-Hydroxy acid2-Hydroxyacetic acid

A701706-25G
77308-160EA 6 USD InStock
77308-160
Glycolic acid 98%
Glycolic acid
Caution: for Research Use Only


Formula: C₂H₄O₃
MW: 76.05 g/mol
MDL Number: MFCD00004312
CAS Number: 79-14-1
UN: 3261
ADR: 8,Ⅱ
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty