Would you like to log in again?
VWR® Basic Protection Beard Cover
Supplier: VWR
VWR®
89522-674
89522-674CS 104.15 USD InStock
89522-674
VWR® Basic Protection Beard Cover
Disposable Garments
VWR® basic protection beard covers are made of non-woven polypropylene material, 11.5 g/m² by weight.
  • Latex-free
  • Non-woven polypropylene
Designed for use in a cleanroom environment. Beard covers are manufactured in a stringent clean environment.
Color: White
Packaging: Beard covers are packaged 100/bag, 10 bags/case.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty