Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Digital Water Bath with Cover, 12 L

Supplier: EDVOTEK
538
470353-790EA 949 USD InStock
470353-790
Digital Water Bath with Cover, 12 L
Baths Boiling Baths
12 L Digital water bath with built-in timer, temperature control, and low-water sensor.

  • Includes large cover and insert covers
  • Temperature range: Ambient to 99.9 °C with cover
  • Digital display
  • Corrosion resistant

The Edvotek® 12 L water bath has a built-in timer. The bath includes digital temperature control and low-water sensor to prevent burn-outs. The unit includes a convenient hose that makes it easy to drain and clean.

Delivery information: Supplied with one cover, as well as four smaller, insert covers.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty