Would you like to log in again?
Atlanta Biomedical Syringe Infusion Pump, FDA Approved
Supplier: Wilburn Medical USA
Atlanta BioMedical 4100
ABC-4100
77296-298EA 5988.89 USD InStock
77296-298
Atlanta Biomedical Syringe Infusion Pump, FDA Approved
Pumps Syringe Pumps
The Atlanta BioMedical Corporation 4100 Syringe Infusion Pump recognizes most IV syringe manufacturers and accepts sizes from 1 ml to 120 ml. US made. Metal housing for durability. Includes a handle and can be mounted on any pole/rail system with an optional pole clamp.
  • Front syringe loading
  • Adjustable flow ranges – changes can be made without interrupting flow
  • Fast bolus rate storage for user defined settings
  • Multiple sensors to monitor delivery accuracy
  • AC or battery operation with 12 hours battery capacity
  • Full alarm capabilities
  • RS232 interface capabilities
  • 1 year parts and labor warranty
Automatic size sensing for 1, 3, 5, 10, 20, 30, 60, and 120 ml syringe sizes.
The ABC-4100 is FDA approved for Intravenous, arterial, epidural, spinal, enteral and subcutaneous delivery routes. Delivery modes include continuous, volume/time, mass, body weight, and bolus. Stores up to 90 customized programmed settings for easy recall.
Certifications: FDA approved.
Accessories information: The ABC rotating pole clamp (sold separately) complements the Model 4100 with its easy IV pole installation and 360° rotation that allows the syringe to face upward. Made in the USA.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty