Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ai EasyVac 2-Stage Vacuum Pump, 9 Cfm with Mist Filter

Supplier: Across International
Ai EasyVac
EV9110220ETL
MSPP-EV9110220EA 972.31 USD InStock
MSPP-EV9110220
Ai EasyVac 2-Stage Vacuum Pump, 9 Cfm with Mist Filter
Pumps Vacuum Pumps
EasyVac 9 CFM vacuum pumps are now 110/220 V 50/60 Hz switchable.

  • Working temperature: −5 to 60 °C
  • Pumping rate: 9 cfm (110 V 60 Hz), 8 cfm (220 V 50/60 Hz)

Ai EV9 are dual-stage high speed sliding-vane rotary compact vacuum pump with KF25 vacuum connector. Comes with all necessary fittings and a exhaust oil mist filter which also returns trapped pump oil back to the pump and makes the pump quieter to run. It is important to note that pumps should always be ran in a well ventilated area.

Certifications: ETL and CE certified.

Delivery information: FreeKF25 flexible stainless steel bellow (3.3"), clamps and centering rings set included.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty