Would you like to log in again?
VWR® Cleanroom Comfort Headbands
Supplier: Avantor
VWR®
37240 37243 37241 37242
77257-034CS 1046.9 USD InStock
77257-034 77257-036 77257-038 77257-040
VWR® Cleanroom Comfort Headbands
Disposable Garments
Sweat absorbing, user-friendly, disposable, and hygienic solution.
  • Premium soft comfort fit
  • Continuous filament fiber stretch nylon
  • Sweat absorbing
  • Cleanroom compatible
  • Disposable
The cleanroom comfort headband is designed for comfort and intended for use under a bouffant hat in cleanroom environments. These headbands are tailor made to ensure user comfort, crafted from continuous filament fiber, stretch nylon for a soft and comfortable cleanroom garment. This product minimizes the risk of contamination and eliminates the need for employees to use external products during business operations.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty