Would you like to log in again?
Triple Horizontal Bars
Supplier: Maze Engineers
5005
76670-190EA 1320 USD InStock
76670-190
Triple Horizontal Bars
Animal Supplies Animal Environmental Enrichment
The triple horizontal bar apparatus is composed of three bars made of stainless steel to assess motor function in rodents.
  • Acrylic frame and stainless steel rods
  • Easy to use and setup
  • Easy to clean, assemble, and disassemble
Customized diameters are available, please inquire for more details.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty