Would you like to log in again?
Circular Active Place Avoidance Arena
Supplier: Maze Engineers
6004
76670-330EA 18653.34 USD InStock
76670-330
Circular Active Place Avoidance Arena
Animal Supplies Animal Environmental Enrichment
The APA is a circular metal arena shock grid underneath a rotating arena to study active and passive avoidance behaviors.
  • Includes grid floor, stainless steel base, rotating arena
  • Shock scrambler, shock cables, high precision serving motor also included
The subjects are allowed to explore the arena while avoiding a prohibited sector by using visual cues in and around the arena. Punishment in the form of shock is delivered to the subject if it enters the prohibited sector. Passive avoidance is observed when the arena is not rotating while active avoidance is observed when the arena is rotating. The arena makes the active place avoidance task suitable for evaluating spatial navigation performances of weanling rodents, making this an effective tool for investigating the effects of brain lesions, pharmacological manipulations, and diseases on spatial learning and memory.
Ordering information: Software package (see accessory option) is designed for use with the automated active place avoidance and video tracking software such as EthoVision or ANY-Maze.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty