Would you like to log in again?
3 Choice Chamber
Supplier: Maze Engineers
4819
76670-124EA 2653.34 USD InStock
76670-124
3 Choice Chamber
Animal Supplies Animal Environmental Enrichment
The three-chamber paradigm is a widely used task to evaluate spatial and non-spatial learning as well as memory in zebrafish.
  • Clear acrylic construction
This watertight apparatus is outlined by a dark panel, covering one side of each compartment as a visual cue to provide an axis of orientation for right/left discrimination. The apparatus is primarily used for learning and memory, but can also be used for various toxicity experiments.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty