Would you like to log in again?
Well Plate Stand, Diversified Biotech
Supplier: Diversified Biotech
WPST-1000
490007-144EA 87.3 USD InStock
490007-144
Well Plate Stand, Diversified Biotech
Well Plate Stands and Well Orienters
A convenient holder for a microplate, tilted at a 30° angle to comfortably view samples and pipette without strain.
Each stand has a neoprene pad and rubber mounting to prevent slippage of the plate and stand.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty