Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

New Brunswick Innova® 40/40R Benchtop Orbital Shakers

Supplier: Eppendorf
New Brunswick Innova®
M1299-0094 M1299-0094 M1299-0080 M1299-0090 M1299-0084 M1299-0090 M1299-0084 M1299-0080
75874-502EA 13411.14 USD InStock
CA75874-408 75874-502 75874-534 CA75875-636 75874-408 75875-636 CA75874-534 CA75874-502
New Brunswick Innova® 40/40R Benchtop Orbital Shakers
Shakers and Mixers Orbital Shakers
The Innova 40/40R is an easy-to-program orbital shaker that comes with the famous Innova triple eccentric drive to provide 24/7 reliable operation for decades.

  • Check your samples: Large, insulated, and scratch-resistent viewing provides a clear view of the chamber contents (device stops when lid is opened); holds flasks up to 3 L
  • Built to perform and last: Innova triple-eccentric counterbalanced drive in cast iron housing provides vibration and trouble-free operation for years
  • Tight temperature and speed control: Temperature accuracy and uniformity: ±0.1 °C and ±0.25 °C (at 37 °C; independent over-temperature cutoff included), speed accuracy 1 rpm
  • Save space on your lab bench: Small footprint of 55.9×76.2 cm (22×30") (W x D) that even works on narrow benches
  • Easy interface handling and programming: Controls and display make it easy to view, set and program parameters - continuous, timed, or multi-step programs with up to 15 steps (four programs storable)
  • Minimizes splashing and mechanical damage: Acceleration/deceleration circuit prevents sudden starts and stops
  • Easy to move: 60/79 kg weight (incubated/refrigerated version)
  • Optional drip pan: Protects the shaker drive below by collecting spills from the overlying universal platform

The Innova 40 and 40R (refrigerated) benchtop shakers provide years of dependable operation. This is the perfect shaker to begin working on small-scale projects with a view to increase volume or throughput. The self-supporting transparent lid allows samples to be in view all the time.

The ability to shake flasks up to 3 L in such a compact benchtop unit provides clear scope to upscale and options include a drip pan and remote alarm. The simple user interface allows for easy programming of speed, temperature, time. The VisioNize box can be added to enhance monitoring and adds notifications functionality. Regular servicing can be carried out in-situ, allowing for minimized downtime.

Memory: Non volatile with automatic power failure restart
Motor type: Solid state, DC brushless motor

Applications:
Grow bacterial, insect, yeast and plant cells in a controlled environment
Screen culture media for optimal growth
Grow seed cultures for fermenters/bioreactors

Certifications: Meets cETL, UL, CSA and CE standards.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty