Would you like to log in again?
Oakton® Flexible Electrode Holder
Supplier: Antylia Scientific
Oakton®
35617-50
76221-268EA 355.54 USD InStock
76221-268
Oakton® Flexible Electrode Holder
Electrode Holders
Keeps electrodes in position.
  • Weighted base pivots 360° for added convenience
Holds up to four standard size electrodes (14 mm dia. max.) and one ATC probe. Weighted base lets you pivot holder 360° for added convenience. Flexible arm lets you move electrode side to side or up and down while keeping electrode at a constant vertical angle.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty