Would you like to log in again?
Idex® Natural FEP Chromatography Tubings
Supplier: Antylia Scientific
Idex®
02015-07 02015-15 02015-16
100463-454EA 266.1 USD InStock
100463-454 100452-402 100452-424
Idex® Natural FEP Chromatography Tubings
Tubing Chromatography Tubing
Excellent chemical resistance at an economical price.
  • Great for moderate to low pressure applications
  • Low gas permeability
FEP (fluorinated ethylene propylene) tubing is ideal choice for general, low pressure applications. Most FEP tubing is translucent, so you can see liquid flow.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty