Would you like to log in again?
Idex® High-Purity PFA Tubings
Supplier: Antylia Scientific
Idex®
02012-70 02012-74
100457-914EA 317.48 USD InStock
100457-914 100452-534
Idex® High-Purity PFA Tubings
Tubing Chromatography Tubing
Manufactured from premium-grade PFA.
  • Low gas permeability and outstanding resistance to solvents
  • Maximum operating temperature 80 °C
High-Purity PFA tubing, manufactured from premium-grade PFA, is one of the most contaminant-free polymers available. It offers high continuous service temperature (recommended max operating temperature 80 °C), smooth surface texture, and excellent polymer purity.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty