Would you like to log in again?
Nichipet Premium LT Single Channel Pipettes
Supplier: Nichiryo
Nichipet®
00-NLT-10 00-NLT-1000 00-NLT-2 00-NLT-20 00-NLT-200 00-NLT-100
10798-802EA 748.86 USD InStock
10798-802 10798-944 10798-800 10798-942 10798-940 10798-938
Nichipet Premium LT Single Channel Pipettes
Pipettes Micropipettes
The Nichipet Premium LT pipette provides innovation in comfort while providing durability and precise results.
  • Mltigates tension to thumb
  • Single hand pipetting can be done smoothly to improve operation efficiency
  • New design for reducing stroke loading and mitigating fatigue
  • Uses an enlarged counter display which is better for visibility and volume setting
  • Seal ring for the Premium is tested dispensing/aspirating 600000 times (2 to 1000 μl)
The Nichipet premium LT provides a hyperblowout system improving dispensing efficiency especially with low volume pipetting. It is easily calibrated in lab, and is fully autoclaveable.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty