Would you like to log in again?
Liquid Weighing Pour Boats
Supplier: Heathrow Scientific
120890V 120891V 120892V
77296-498CS 105.84 USD InStock
77296-498 77296-496 77296-494
Liquid Weighing Pour Boats
Boats Weighing Boats
Convenient and specific tool for customers who need to weigh liquids on a scale. Deep vessel features pour spout and sturdy handle for safe and effective transport and pouring.
  • Flat bottom ensures stability and prevents tipping
  • Operating temperature range 0 to 40 °C (104 °F)
  • Made from FDA food grade polystyrene (0.45 mm thick) in an ISO Class 8 clean room
  • Recyclable, marked
Material biologically inert and resistant to dilute and weak acids, aqueous solutions, alcohols and bases.
Available in 3 sizes (50, 100 and 200 ml).
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty