Would you like to log in again?
MyFuge™ Mini Microcentrifuge
Supplier: Alkali Scientific
MyFuge™
C9310
104028-684EA 332.29 USD InStock
104028-684
MyFuge™ Mini Microcentrifuge
Centrifuges Microcentrifuges
The MyFuge™ mini centrifuge is a personal centrifuge that fits in the palm of your hand. Ideal for quick spin downs of microtubes and PCR tubes, the MyFuge mini is extremely easy to use. Simply close the lid and the rotor instantly reaches 6000 rpm. Open the lid, and the rotor quickly decelerates to a stop for removing samples. An increased capacity, eight-position microtube rotor is included and comes installed.
  • Eight position microtube rotor
  • Four position PCR strip rotor
  • Storage compartment for PCR rotor
  • Near silent operation
  • Starts and stops in seconds
  • Conserves valuable bench space
Also included is a second rotor for 0.2 ml PCR tubes and strips which is stored (together with the rotor removal key) in a convenient compartment on the bottom of the centrifuge. When the rotor is not in use, storage in this compartment prevents loss or misplacement of the rotor. The MyFuge™ Min is compatible with Quick-Snap™ 1.5 ml microcentrifuge tubes and more.
Ordering information: With 2 years warranty.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty