Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Water, GemPure™, Cell and Molecular Grade

Supplier: GeminiBio
Synonyms: Dihydrogen oxide

901-007-C23 901-007-C23
77322-352EA 202.97 USD InStock
CA77322-352 77322-352
Water, GemPure™, Cell and Molecular Grade
Water
GeminiBio’s Cell and Molecular Grade Water is manufactured specifically for your most stringent research applications. GemPure™ Cell and Molecular Grade Water is sterile and low endotoxin for your cell culture processes. Protease and nuclease free, it is perfect for any of your molecular biology needs as well saving valuable lab space and simplifying your purchasing process.

  • Tested for RNase, DNase, Protease, sterility, TOC, endotoxin, and more
  • Manufactured in an ISO13485:2016 and 21CFR820 compliant facility
  • Available in 1L bottle and custom sizes to suit your needs
  • For further manufacturing or research use only
  • Not for direct Therapeutic use


Formula: H₂O
MW: 18.02 g/mol
MDL Number: MFCD00011332
CAS Number: 7732-18-5
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty