Would you like to log in again?
Hermle Z287-A Compact Universal Centrifuge, 230 V
Supplier: Ward's
Z287-A-E
470334-122EA 4775 USD InStock
470334-122
Hermle Z287-A Compact Universal Centrifuge, 230 V
Centrifuges Benchtop Centrifuges
The Hermle Z287-A Compact Universal Centrifuge exceeds the limitations set forth by traditional microcentrifuges which are restricted solely to fixed angle rotors for microtubes. Instead, this compact, next generation model features an expansive line of swing out and fixed angle rotors options that accept microplates and test-tubes up to 50 ml. This centrifuge is designed to be our most compact and economical. It is a true workhorse for any lab requiring a centrifuge that can accommodate many fields of research.
  • 230 V model, not for use in USA
  • Automatic rotor identification
  • Electrical safety lid-lock
  • Momentary spin button
  • Microprocessor Control
  • Over-speed protection
  • Adjustable Accel/Decel
The Z287-A introduces a convenient status indicator. Users can easily view the current state of the centrifuge from across the room. The LED illuminates to signify if the rotor is spinning, the run is completed or the centrifuge is idle in standby mode.
Advanced microprocessor controls allow for instant rotor identification, which automatically converts the rotational speed to the relevant g-force (rcf), preventing the user from setting a speed above the capabilities of the installed rotor. In addition to retrieving saved programs, the EZ-Scroll touch wheel front panel can adjust accel/decel rates, and change the end of run alarm tone. Additionally this feature prevents debris buildup, leakage, and can be operated with gloves on. The Z287-A is a truly powerful and versatile centrifuge perfect for any research laboratory.
Power: 170 W, timer: Up to 99 hours, 59 minutes.
Caution: 230 V model, not for use in USA. Corresponding model with voltage suitable for use in US (470334-120) is also available, please see separate product page.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty