Would you like to log in again?
Sprint™ Clinical Centrifuges, 230 V
Supplier: Ward's
C5000-6H-E C3303-6HP-E
470329-986EA 609 USD InStock
470329-986 470329-990
Sprint™ Clinical Centrifuges, 230 V
Centrifuges Benchtop Centrifuges
These economical swing out rotor centrifuges provide fixed-speed or variable speed capabilities for all your blood and urine collection tubes up to 15 ml.
  • 230 V models, not for use in USA
  • 6 position swing-out rotor
  • Choose either fixed or variable speed
  • Adjustable timer
These clinical centrifuges are the ideal economical choice for all your blood, urine and cell separation needs. Both models offer a variable timer and digital display for settings. The basic model has a fixed speed, while the Plus model offers variable speed, in addition to a larger capacity tube (up to 15 ml, round or conical bottom), as well as increased speed, a safety imbalance detection system, momentary operation, and adjustable acceleration and deceleration levels. Both come with a 2 year warranty.
Certifications: UL listed.
Caution: 230 V models, not for use in USA. Corresponding models with voltage suitable for use in US (470329-984 and 470329-988) are also available, please see separate product page.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty