Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Methyl-2-(methylthio)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

108113-500MG 108113-5G 108113-1G
77371-502EA 3132 USD InStock
77371-502 77339-800 77347-498
5-Methyl-2-(methylthio)benzaldehyde ≥97%
5-Methyl-2-(methylthio)benzaldehyde
Caution: for Research Use Only


Formula: C₉H₁₀OS
MW: 166.24 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24674059
CAS Number: 13616-73-4
Order Now


 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty