Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-(2-Bromo-6-chlorophenyl)-2-methylpropan-1-ol ≥97%

Supplier: AOBChem USA

Warning

117446-5G 117446-25G 117446-500MG 117446-1G
77347-088EA 248 USD InStock
77347-088 77359-734 77383-570 77323-606
1-(2-Bromo-6-chlorophenyl)-2-methylpropan-1-ol ≥97%
1-(2-Bromo-6-chlorophenyl)-2-methylpropan-1-ol
Caution: for Research Use Only


Formula: C₁₀H₁₂BrClO
MW: 263.559 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30157658
CAS Number: 1878366-07-4
Order Now


 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty