Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Bromo-2-(methylsulfonyl)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

122211-5G 122211-25G 122211-1G 122211-500MG
77347-780EA 405 USD InStock
77347-780 77371-876 77323-732 77383-656
5-Bromo-2-(methylsulfonyl)benzoic acid ≥97%
5-Bromo-2-(methylsulfonyl)benzoic acid
Caution: for Research Use Only


Formula: C₈H₇BrO₄S
MW: 279.10 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23701693
CAS Number: 146431-33-6
Order Now


 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty