Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Chloro-6-fluorobenzaldehyde oxime ≥97%

Supplier: AOBChem USA

Warning
Synonyms: 2-Chloro-6-fluorobenzaldoxime

31037-25G 31037-5G
77373-222EA 207 USD InStock
77373-222 77361-544
2-Chloro-6-fluorobenzaldehyde oxime ≥97%
2-Chloro-6-fluorobenzaldehyde oxime
Caution: for Research Use Only


Formula: C₇H₅ClFNO
MW: 173.57 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00837704
CAS Number: 443-33-4
Order Now


 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty