Would you like to log in again?
Elite Series Waterproof ORP Pocket Tester, Thermo Scientific
Supplier: Thermo Scientific
Elite
ELITEORP
76284-656EA 275.83 USD InStock
76284-656
Elite Series Waterproof ORP Pocket Tester, Thermo Scientific
Electrochemical Meters pH and Multi-Parameter Meters
Elite Pocket Testers offer flexible on-the-go electrochemical testing. Choose the Elite ORP model to measure oxidation-reduction potential (ORP) using a direct-measure sensor design for your application.
  • Large back-lit graphic display
  • Uses 4 x AAA alkaline batteries
  • Average 150 hours of battery life
  • Wide leak-proof cap serves as stable base
  • Replaceable sensors
Ideal for testing beverages including homebrew beer and wine, food, hydroponics, aquaculture and aquariums, environmental monitoring and more.
One-year warranty for testers. Six-month warranty for replacement sensors.
Certifications: Meets IP67 waterproof rating.
Ordering information: Supplied with batteries, lanyard and soft carry case. Replacement sensor can be ordered separately.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty