Would you like to log in again?
Compact Centrifuges 5702, 230 V, Eppendorf
Supplier: Eppendorf
022629916 022629888 022626256 022626108 022626230
80081-164EA 3904.42 USD InStock
80081-164 80081-166 80076-420 89203-054 75875-104
Compact Centrifuges 5702, 230 V, Eppendorf
Centrifuges Benchtop Centrifuges
These compact, low-speed centrifuges can accommodate tube volumes from 1.1 to 85 ml. They are ideal for tissue culture, blood separations, phenol chloroform extractions (Phase Lock Gel), cell harvesting, and standard low-speed applications. Features include a brushless induction motor; digital display of rpm, RCF, and time; rpm to RCF conversion; a separate short spin button; and audible alarm at end of run. Time and speed parameters can be locked to avoid unintentional changes from occurring. Quick acceleration and deceleration times minimize waiting, while the lid opens automatically at the end of a run for convenient, quick access.
  • 230 V models, not for use in USA
  • Centrifuge 5702 for standard applications
  • Refrigerated Centrifuge 5702 R for heat sensitive samples
  • Heated and refrigerated Centrifuge 5702 RH allows centrifugation to improve cell viability for more accurate results in downstream applications
In addition to these features, fast cool function that eliminates long waiting times
Refrigerated-heated model 5702RH possesses the features of model 5702R as well as the ability to accurately maintain temperatures higher than room temperature. Temperature-sensitive cells or cell cultures can be centrifuged under defined conditions, such as 37 °C. Temperature deviation in stationary state at 4 °C and at 37 °C is ≤1 °C.
Clinical Packages and Cell Culture Packages include centrifuge, rotor, buckets, and adapters for vacutainer tubes. Non-Clinical Packaged Models 5702, 5702R, and 5702RH use the same rotors, which must be ordered separately (see Rotors and Accessories for Eppendorf® Compact Centrifuges).
Model 5702R also has two blue-lit program buttons for quick and easy recall of frequently used parameters, as well as two timer modes: standard and “at set rpm.” The standard mode begins counting when the start button is pressed and the “at set rpm” timer begins when the centrifuge reaches nominal speed, providing more consistent separation results.
Ordering information: Supplied with a manufacturer’s two-year parts and labor warranty
Caution: 230 V models, not for use in USA. Corresponding models with voltage suitable for use in US are also available, please see separate product page.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty