Would you like to log in again?
Thermo Scientific™ NanoDrop™ Lite Plus Spectrophotometers
Supplier: Thermo Fisher Scientific
NanoDrop™
NDLPLUSPRGL
MSPP-NDLPLUSPREA 12307.69 USD InStock
MSPP-NDLPLUSPR
Thermo Scientific™ NanoDrop™ Lite Plus Spectrophotometers
Spectrophotometers and Photometers Biophotometers
Quickly and easily assess sample concentration and purity ratios with the Thermo Scientific™ NanoDrop™ Lite Plus Microvolume Spectrophotometer, the entry-level complement to the full-featured Thermo Scientific™ NanoDrop™ One/OneC and NanoDrop Eight Spectrophotometers. While offering fewer features than our premium models, this instrument delivers fast, accurate measurements in an easy-to-use, affordable package.
  • Economical
  • Standard measurements
  • Fast, accurate measurements from only 1 to 2 μl saving precious samples, no waste
  • Built-in touchscreen operation (no computer required)
  • Data storage and transfer, last 1000 measurements saved locally; export data to a USB device
  • Optional printer available for freezer-compatible labels (not included)
  • Compact and portable—fits on any lab bench, ready for your next experiment
The NanoDrop Lite Plus Spectrophotometer is a valuable teaching tool and is ideal for quality check measurements in busy research labs as well. Measure purified DNA, RNA, and protein concentration up to 30 Abs and calculate critical A260/A280 and A260/A230 purity ratios using trusted NanoDrop technology.
The NanoDrop Lite Plus Spectrophotometer is designed for routine measurements of nucleic acid and protein samples. The sample-retention technology eliminates the need to dilute highly concentrated samples, making measurements easy. Simply pipette your sample directly onto the optical measurement pedestal, measure, and wipe clean with a lint-free wipe. Its compact design, with built-in controls and software, means the NanoDrop Lite Plus Spectrophotometer takes up little space on your benchtop.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty