Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
DC-Regulated Fiber Optic Illuminator System, Cole Parmer
Supplier: Cole Parmer
EW-41720-40
MSPP-EW4172040EA 1593.46 USD InStock
MSPP-EW4172040
DC-Regulated Fiber Optic Illuminator System, Cole Parmer
Lights Halogen Lights, Lamps and Bulbs
Avoid lighting problems due to variation in input voltage.
  • Ideal for video microscopy and machine vision applications
  • One year warranty
The internal power supplyin the illuminator to the halogen bulb is stabilized to ±0.5%, eliminating problems due to variation in input line voltages. Front-panel dial controls light output from 0 to 100%. Remotely control the light output using a 0 to 5 VDC signal. Stackable housing is heavy-duty steel. Includes a 150 W bulb and 6' power cord with three-prong plug.
Illuminator dimension: 18.41W×24.13D×11.75H cm (7.25×9.5×4.6")
Certifications: UL/cUL listed, CE approved, and RoHS compliant.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty