Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Eppendorf® Multipurpose Centrifuges 5804, 5804R, 5810 and 5810R, 220/230 V

Supplier: Eppendorf
022622552 022623559 022623559 022625055 022625551 022622552
53513-810EA 11238.18 USD InStock
53513-810 53513-802 53513-814 53513-806 CA53513-802 CA53513-806
Eppendorf® Multipurpose Centrifuges 5804, 5804R, 5810 and 5810R, 220/230 V
Centrifuges Benchtop Centrifuges
Eppendorf® centrifuges 5804, 5804R, 5810, and 5810R, with their renowned quality and reliability, offer the most cost efficient solutions for medium to high-throughput applications.

  • 220/230 V models, not for use in USA
  • Powerful and maintenance-free motors
  • Accommodate a variety of rotors for unparalleled application versatility

They are designed to fit in every lab, the 5804, 5804R, 5810, and 5810R feature a compact footprint, which saves valuable bench space

They are three centrifuges in one: a high capacity general purpose centrifuge for cell harvesting; a high-speed centrifuge for separating cell lysates; and a micro centrifuge for DNA precipitations. Interchangeable swing-bucket and high-speed fixed-angle rotors spin microcentrifuge tubes, popular glass and plastic tubes, microplates, and blood collection tubes.

Centrifuges also accommodate fixed-angle and swing-bucket rotors for a variety of applications. A large selection of adapters allows maximum application flexibility with a minimum number of rotors.

Refrigerated centrifuge 5804R and 5810R features outstanding software control, which guarantees highly accurate timing (±2 s in 5 min.), speed (1%), and temperature (±2 °C at 6 °C ) for maximum run reproducibility. Temperature range from –9 to 40 °C with FastTemp function for rapid cooling of the centrifuge chamber, and standby cooling, which maintains temperature when not in use, are standard features.

Certifications: UL and C-UL listed. CSA certified.

Ordering information: Available packages include Cell Culture packages, with a swing-bucket rotor and adapters for standard 15 and 50 ml conical tubes, and Clinical packages, with a swing-bucket rotor and adapters for applications in standard 13 and 16 mm blood collection or urine tubes.

Caution: 220/230 V models, not for use in USA. Corresponding models with voltage suitable for use in US are also available, please see separate product page.

Order Now
 

ORDER

SPECIFICATIONS

ACCESSORIES

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty