Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Corning® Elplasia® 12K Open Well Plate

Supplier: Corning
Corning® Elplasia® 12K
4547 4547
77135-260CS 403.89 USD InStock
CA77135-260 77135-260
Corning® Elplasia® 12K Open Well Plate
Microplates Cell Culture Plates
Culture thousands of spheroids in a microplate footprint.

  • Ease of spheroid formation, culture, assessment, and harvest
  • Ready-to-use plate to create large quantities of uniform spheroids
  • Scaffold-free cultures
  • Culture spheroids for up to 14 or more days (cell line-dependent)
  • Highly reproducible bulk spheroid formation across microcavities
  • Common media reservoir for equivalent culture conditions for all spheroids
  • Easy access to spheroid cultures
  • Compatible with brightfield and fluorescent microscopy

Critical in drug screening and other applications such as cell therapy research, spheroids are coveted for their ability to more closely resemble the original tumor from which they were derived. The Corning Elplasia 12K plate is designed similarly to the Corning Elplasia 12K flask, but in a microplate footprint which makes it ideal for easy sampling and imaging of spheroids. The plate also features media exchange ports for ease of liquid handling.

No. of microcavities: 12000
Microcavity dimensions: 500W×600 µm D

Certifications: Sterile.

Packaging: 1/pack, 5/case.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty